Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EPHA5 Rabbit polyclonal antibody FITC (Fluorescein)

Dodávateľ: US Biological
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: EPHA5 (EPH receptor A5)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
200 µl 200 µl USBIE3363-01F-FITC
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF

ŠPECIFIKÁCIE

Antigen symbol EPHA5
Antigen name EPH receptor A5
Reactivity Human, Mouse
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitope C-terminal
Purification purified by Protein G chromatography
Form liquid
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Storage temperature Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo