Tlačiť
Anti-EPHA5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Dodávateľ: United States Biological
USBIE3363-01F-PEEA 947.16 EUR
USBIE3363-01F-PE
Anti-EPHA5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: EPHA5 (EPH receptor A5)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse