Tlačiť

Anti-EPHA6 Rat monoclonal antibody [clone: 9G64]

USBIE3363-05
Anti-EPHA6 Rat monoclonal antibody [clone: 9G64]
Type: Primary
Antigen: EPHA6 (EPH receptor A6)
Clonality: monoclonal
Clone: 9G64
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR