Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-EPHA7 Rat monoclonal antibody unconjugated [clone: 9G65]

Dodávateľ: US Biological
Type: Primary
Antigen: EPHA7 (EPH receptor A7)
Clonality: monoclonal
Clone: 9G65
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse



Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG USBIE3363-07B
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol EPHA7
Antigen name EPH receptor A7
Reactivity Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 9G65
Host Rat
Isotype IgG1
Western Blot Yes
Purification purified
Form lyophilized powder
Storage Buffer PBS
Storage temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo