Tlačiť

Anti-EPHA7 Mouse Monoclonal Antibody [clone: 6C8G7]

Dodávateľ: United States Biological
USBIE3363-07CEA 703.86 EUR
USBIE3363-07C
Anti-EPHA7 Mouse Monoclonal Antibody [clone: 6C8G7]
Protilátky
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200

Type: Primary
Antigen: EPHA7 (EPH receptor A7)
Clonality: monoclonal
Clone: 6C8G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR