Tlačiť

Anti-EPHA10 Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: United States Biological
USBIE3363-13A-BIOTEA 949.22 EUR
USBIE3363-13A-BIOT
Anti-EPHA10 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: EPHA10 (EPH receptor A10)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR