Tlačiť

Anti-EPHA10 Rabbit Polyclonal Antibody

Dodávateľ: United States Biological
USBIE3363-13BEA 625.17 EUR
USBIE3363-13B
Anti-EPHA10 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: EPHA10 (EPH receptor A10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR