Tlačiť

Anti-EPHA10 Mouse Monoclonal Antibody [clone: 11n65]

Dodávateľ: United States Biological
USBIE3363-13CEA 845.82 EUR
USBIE3363-13C
Anti-EPHA10 Mouse Monoclonal Antibody [clone: 11n65]
Protilátky
Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: EPHA10 (EPH receptor A10)
Clonality: monoclonal
Clone: 11n65
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR