Tlačiť
Anti-Histone H4, trimethyl Rabbit monoclonal antibody [clone: D84D2]
Dodávateľ: United States Biological
USBIH5110-17MEA 678.52 EUR
USBIH5110-17M
Anti-Histone H4, trimethyl Rabbit monoclonal antibody [clone: D84D2]
Protilátky
Recommended dilution: Western Blot: 1:1000ChImmunoprecipitation: 1:50
Type: Primary
Antigen: Histone H4, trimethyl
Clonality: monoclonal
Clone: D84D2
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Monkey, Mouse, Pig, Rat, Xenopus laevis