Tlačiť

Anti-IL8 Rabbit Polyclonal Antibody

Dodávateľ: United States Biological
USBII8430-03LEA 621.62 EUR
USBII8430-03L
Anti-IL8 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 0.5-2?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: IL8 (interleukin 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR