Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-IL8 Rat monoclonal antibody unconjugated [clone: 9H141]

Dodávateľ: US Biological
Recommended dilution: ELISA: 2-8μg/ml

Type: Primary
Antigen: IL8 (interleukin 8)
Clonality: monoclonal
Clone: 9H141
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Cat



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
500 µg 500 µG USBII8430-22C
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF

ŠPECIFIKÁCIE

Antigen symbol IL8
Antigen name interleukin 8
Reactivity Cat
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 9H141
Host Rat
Isotype IgG2a
ELISA Yes
Neutralisation Yes
Amino acid number 23 to 101
Concentration ~500 ug/ml
Purification purified by Protein G chromatography
Form lyophilized powder
Storage buffer 0,2um sterile-filtered solution, PBS, 5% trehalose.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo