Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-IL8 Mouse monoclonal antibody NL®637 [clone: 9j5]

Dodávateľ: US Biological
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: IL8 (interleukin 8)
Clonality: monoclonal
Clone: 9j5
Conjugation: NL®637
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
500 µl 500 µl USBII8430-26W
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF

ŠPECIFIKÁCIE

Antigen symbol IL8
Antigen name interleukin 8
Reactivity Human
Conjugation NL® 637
Clonality Monoclonal
Antibody type Primary
Clone 9j5
Host Mouse
Isotype IgG1
ImmunoChemistry Yes
Amino acid number 28 to 99
Purification purified by Protein G chromatography
Form liquid
Storage buffer PBS, 0,1% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo