Tlačiť

Anti-Interleukin 8 Receptor B Mouse Monoclonal Antibody [clone: 6D499]

Dodávateľ: United States Biological
USBII8430-35FEA 511.44 EUR
USBII8430-35F
Anti-Interleukin 8 Receptor B Mouse Monoclonal Antibody [clone: 6D499]
Protilátky
Type: Primary
Antigen: Interleukin 8 Receptor B
Clonality: monoclonal
Clone: 6D499
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR