Tlačiť

Anti-IL9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K114]

Dodávateľ: United States Biological
USBII8431-04JEA 826.71 EUR
USBII8431-04J
Anti-IL9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K114]
Protilátky
Do not freeze!

Type: Primary
Antigen: IL9 (interleukin 9)
Clonality: monoclonal
Clone: 12K114
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR