Tlačiť

Anti-IL9R Mouse Monoclonal Antibody [clone: 8H148]

Dodávateľ: United States Biological
USBII8431-75NEA 1065.06 EUR
USBII8431-75N
Anti-IL9R Mouse Monoclonal Antibody [clone: 8H148]
Protilátky
Type: Primary
Antigen: IL9R (interleukin 9 receptor)
Clonality: monoclonal
Clone: 8H148
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR