Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-IL9R Mouse monoclonal antibody PE (Phycoerythrin) [clone: 8H148]

Dodávateľ: US Biological
Do not freeze!

Type: Primary
Antigen: IL9R (interleukin 9 receptor)
Clonality: monoclonal
Clone: 8H148
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 Tests 100 Tests USBII8431-75P
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF

ŠPECIFIKÁCIE

Antigen symbol IL9R
Antigen name interleukin 9 receptor
Reactivity Human
Conjugation PE (Phycoerythrin)
Clonality Monoclonal
Antibody type Primary
Clone 8H148
Host Mouse
Isotype IgG1
Flow cytometry Yes
Amino acid number 40 to 270
Concentration ~0,025 mg/ml
Purification purified by Protein G chromatography
Form liquid
Storage buffer saline, 0,5% BSA, 0,1% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo