Tlačiť

Anti-IL10 Mouse Monoclonal Antibody [clone: 5J48]

Dodávateľ: United States Biological
USBII8432-01FEA 864.65 EUR
USBII8432-01F
Anti-IL10 Mouse Monoclonal Antibody [clone: 5J48]
Protilátky
Recommended dilution: ELISA: 1-5 ?g/ml

Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 5J48
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR