Tlačiť

Anti-IL10 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6D63]

Dodávateľ: United States Biological
USBII8432-01H1EA 627.15 EUR
USBII8432-01H1
Anti-IL10 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6D63]
Protilátky
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 6D63
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR