Tlačiť

Anti-IL10 Mouse Monoclonal Antibody [clone: 5J17]

Dodávateľ: United States Biological
USBII8432-01MEA 772.48 EUR
USBII8432-01M
Anti-IL10 Mouse Monoclonal Antibody [clone: 5J17]
Protilátky
Recommended dilution: Western Blot/DB: 01-1 ?g

Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 5J17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR