Tlačiť

Anti-IL10 Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: United States Biological
USBII8432-02D1EA 744.12 EUR
USBII8432-02D1
Anti-IL10 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 0.25-1 ?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR