Tlačiť

Anti-IL10 Rat Monoclonal Antibody [clone: 10B2358]

Dodávateľ: United States Biological
USBII8432-02GEA 613.56 EUR
USBII8432-02G
Anti-IL10 Rat Monoclonal Antibody [clone: 10B2358]
Protilátky
Recommended dilution: Flow Cytometry: 1:10-1:100; ; Western Blot: 1:500-1:1000

Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 10B2358
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR