Tlačiť

Anti-IL10 Sheep Polyclonal Antibody

Dodávateľ: United States Biological
USBII8432-11DEA 896.78 EUR
USBII8432-11D
Anti-IL10 Sheep Polyclonal Antibody
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR