Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-IL10 Rat monoclonal antibody unconjugated [clone: 5K315]

Dodávateľ: US Biological
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 5K315
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Mouse



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
500 µg 500 µG USBII8432-18A
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF

ŠPECIFIKÁCIE

Antigen symbol IL10
Antigen name Interleukin 10
Reactivity Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 5K315
Host Rat
Isotype IgG1 kappa
Western blot Yes
ELISA Yes
Concentration ~0,5 mg/ml
Purification purified
Form liquid
Storage buffer 100mM borate buffered saline, pH 8.0. No oramine- containing buffer salts .
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo