Tlačiť
Anti-IL10 Rat Monoclonal Antibody (Biotin) [clone: 5K315]
Dodávateľ: United States Biological
USBII8432-18BEA 527.17 EUR
USBII8432-18B
Anti-IL10 Rat Monoclonal Antibody (Biotin) [clone: 5K315]
Protilátky
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 5K315
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Mouse