Tlačiť

Anti-IL10 Rat Monoclonal Antibody [clone: 5J337]

Dodávateľ: United States Biological
USBII8432-23CEA 601.34 EUR
USBII8432-23C
Anti-IL10 Rat Monoclonal Antibody [clone: 5J337]
Protilátky
Recommended dilution: ELISA: 1-5 ?g/ml

Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 5J337
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR