Tlačiť

Anti-IL10 Rat Monoclonal Antibody (Biotin) [clone: 5K316]

Dodávateľ: United States Biological
USBII8432-24CEA 526.03 EUR
USBII8432-24C
Anti-IL10 Rat Monoclonal Antibody (Biotin) [clone: 5K316]
Protilátky
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 5K316
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR