Tlačiť

Anti-IL10 Rat Monoclonal Antibody [clone: 5K303]

Dodávateľ: United States Biological
USBII8432-25AXEA 533.52 EUR
USBII8432-25AX
Anti-IL10 Rat Monoclonal Antibody [clone: 5K303]
Protilátky
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: 5K303
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR