Tlačiť

Anti-Interleukin 10 Receptor alpha Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10B2192]

Dodávateľ: United States Biological
USBII8432-28A3EA 447.16 EUR
USBII8432-28A3
Anti-Interleukin 10 Receptor alpha Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10B2192]
Protilátky
Do not freeze!

Type: Primary
Antigen: Interleukin 10 Receptor alpha
Clonality: monoclonal
Clone: 10B2192
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR