Tlačiť

Anti-Interleukin 10 Receptor alpha Rat Monoclonal Antibody (Biotin) [clone: 10B2190]

Dodávateľ: United States Biological
USBII8432-28B2EA 576.17 EUR
USBII8432-28B2
Anti-Interleukin 10 Receptor alpha Rat Monoclonal Antibody (Biotin) [clone: 10B2190]
Protilátky
Recommended dilution: Flow Cytometry:1:10;

Type: Primary
Antigen: Interleukin 10 Receptor alpha
Clonality: monoclonal
Clone: 10B2190
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR