Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-Interleukin 10 Receptor alpha Rat monoclonal antibody PE (Phycoerythrin) [clone: 10B2190]

Dodávateľ: US Biological
Do not freeze!

Type: Primary
Antigen: Interleukin 10 Receptor alpha
Clonality: monoclonal
Clone: 10B2190
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human



 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 Tests 100 Tests USBII8432-28B3
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF

ŠPECIFIKÁCIE

Antigen symbol IL10RA
Antigen name Interleukin 10 Receptor alpha
Reactivity Human
Conjugation PE (Phycoerythrin)
Clonality Monoclonal
Antibody type Primary
Clone 10B2190
Host Rat
Isotype IgG2a
Flow cytometry Yes
Purification purified by Protein G chromatography
Form lyophilized powder
Storage buffer PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo