Tlačiť
Anti-MAGEA2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Dodávateľ: United States Biological
USBIM1750-03-APCEA 947.16 EUR
USBIM1750-03-APC
Anti-MAGEA2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: MAGEA2 (melanoma antigen family A, 2)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human