Tlačiť

Anti-Protein Phosphatase 6 Rabbit Polyclonal Antibody

Dodávateľ: United States Biological
USBIP9107-56KEA 703.86 EUR
USBIP9107-56K
Anti-Protein Phosphatase 6 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: Protein Phosphatase 6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR