Tlačiť

Anti-PPT1 Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: United States Biological
USBIP9107-90-BIOTEA 949.22 EUR
USBIP9107-90-BIOT
Anti-PPT1 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: PPT1 (palmitoyl-protein thioesterase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR