Chcete sa opäť prihlásiť?
Anti-Stage Specific Embryonic Antigen 4 Mouse monoclonal antibody unconjugated [clone: 8H218]
Dodávateľ: US Biological
Type: Primary
Antigen: Stage Specific Embryonic Antigen 4
Clonality: Monoclonal
Clone: 8H218
Conjugation: Unconjugated
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
Rozšíriť všetko / Schovať všetko
Veľkosť Pk Číslo v katalógu VWR Množstvo
100 µg 100 µG USBIS6610-11
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Antigen symbol SSEA-4
Antigen name Stage Specific Embryonic Antigen 4
Reactivity Human, Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 8H218
Host Mouse
Isotype IgG3
Flow cytometry Yes
Concentration ~0,5 mg/ml
Purification purified by Protein A chromatography
Form lyophilized powder
Storage buffer TBS,5% trehalose.
Storage temperature Cool
Shipping temperature Ice
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo