Tlačiť
Goat Anti-Rabbit IgG F(c) Polyclonal Antibody (DyLight® 800)
IMMRIR2180
Goat Anti-Rabbit IgG F(c) Polyclonal Antibody (DyLight® 800)
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit