Tlačiť
Plate washer, PROTEAN II XI
BRAD165-1991
Plate washer, PROTEAN II XI
Plate washer includes two plate holders, washing tank, lid, and one bottle of cleaning concentrate. Accomodates up to 8 plates.