Tlačiť

Anti-IgG F(c) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

AGRIAS10810EA 0 EUR
AGRIAS10810
Anti-IgG F(c) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR