CF™660C and CF™660R antibodies are affinity-purified antibodies labeled with far-red fluorescent CF™660C or CF™660R dyes, members of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™660C is a far-red fluorescent dye with excitation/emission maxima at 667/685nm. CF™660C is several fold brighter and more photostable than the spectrally similar dye AlexaFluor®660. CF™660R is far-red fluorescent dye with excitation/emission maxima at 663/682nm. CF™660R is also brighter than AlexaFluor®660, and is the most photostable 660nm dye available, making it a superior choice for demanding applications like confocal microscopy.
CF™ dyes are covered by pending U.S. and international patents. CF™ is a trademark of Biotium, Inc. Alexa Fluor® is a registered trademark of Molecular Probes, Inc.
Objednávacie informácie: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF660C
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit