Tlačiť
Anti-STRA8 Mouse Monoclonal Antibody [clone: 526CT11.1.5]
Dodávateľ: Abgent
ABGEAM2091AEA 386.83 EUR
ABGEAM2091A
Anti-STRA8 Mouse Monoclonal Antibody [clone: 526CT11.1.5]
Protilátky
Type: Primary
Antigen: STRA8
Clonality: Monoclonal
Clone: 526CT11.1.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse