Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-TAF8 Rabbit Polyclonal Antibody

Dodávateľ: Abgent
Type: Primary
Antigen: TAF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human




 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Pk Číslo v katalógu VWR Množstvo
400 µl ABGEAP13397C
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol TAF8
Antigen name TAF8 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 43kDa
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms TAFII43, TBN, II, TAF, TAFII-43
Amino acid number 151 to 178
Gene ID 129685
Epitope internal
Concentration 0.73
Immunogen This TAF8 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 151-178 amino acids from the Central region of human TAF8.
Molecular weight 34.262 kDa
Purification Protein A & Peptide affinity purification
Sequence REPVSDYQVLREKC
Storage buffer pH 7,4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo
 

Nový vzhľad a obnovený záväzok

Postavené na funkciách a zdrojoch, ktoré poznáte, optimalizované s inováciami od spoločnosti Avantor®. Naša nová značka symbolizuje agilitu a našu schopnosť prispôsobiť sa potrebám zákazníkov.

ĎALŠIE INFORMÁCIE ZAČAŤ NAKUPOVAŤ