Tlačiť

Anti-ABI3BP Rabbit Polyclonal Antibody

Dodávateľ: Abgent
ABGEAP19014CEA 386.83 EUR
ABGEAP19014C
Anti-ABI3BP Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ABI3BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR