Tlačiť
Anti-DPPA2 Mouse Monoclonal Antibody [clone: 210CT5.14.6.1]
Dodávateľ: Abgent
ABGEAM1915BEA 387.67 EUR
ABGEAM1915B
Anti-DPPA2 Mouse Monoclonal Antibody [clone: 210CT5.14.6.1]
Protilátky
Type: Primary
Antigen: DPPA2
Clonality: Monoclonal
Clone: 210CT5.14.6.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human