Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-ACTB1 Rabbit Polyclonal Antibody

Dodávateľ: Abgent
Type: Primary
Antigen: ACTB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Zebrafish




Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Pk Číslo v katalógu VWR Množstvo
400 µl ABGEAZB10021C
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol ACTB1
Antigen name actin, beta 1
Reactivity Zebrafish
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms ACTB|B-ACTZF|actba|bact|bactin1|bactzf|wu:fd18f05
Amino acid number 188 to 215
Gene ID 57934
Epitope internal
Concentration 0.5
Immunogen This DANRE actba antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 188-215 amino acids from the Central region of DANRE actba.
Molecular weight 41.767 kDa
Purification Protein A & Peptide affinity purification
Sequence TAEREIVRDIKEKC
Storage buffer pH 7,4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo