Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Anti-ST18 Rabbit Polyclonal Antibody

Dodávateľ: Abgent
Type: Primary
Antigen: ST18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse



Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
 

OBJEDNAŤ

Rozšíriť všetko / Schovať všetko
Pk Číslo v katalógu VWR Množstvo
400 µl ABGEAP19382A
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Antigen symbol ST18
Antigen name Suppression of Tumorigenicity 18
Reactivity Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms Myt3, Nzf3, NZF-3, AV348974, mKIAA0535
Amino acid number 195 to 222
Gene ID 240690
Epitope N-terminal
Concentration 0.88
Immunogen This Mouse St18 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 195-222 amino acids from the N-terminal region of mouse St18.
Molecular weight 114.75 kDa
Purification Protein A & Peptide affinity purification
Sequence SEDTDTHKGPKHKC
Storage buffer pH 7,4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo