Tlačiť

Anti-DAPK1 Rabbit Polyclonal Antibody

Dodávateľ: Abgent
ABGEAP16359AEA 387.67 EUR
ABGEAP16359A
Anti-DAPK1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DAPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR