Tlačiť

Anti-PRR5 Rabbit Polyclonal Antibody

Dodávateľ: Abgent
ABGEAP19341BEA 386.83 EUR
ABGEAP19341B
Anti-PRR5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PRR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR