Tlačiť

Anti-RALYL Rabbit Polyclonal Antibody

Dodávateľ: Abgent
ABGEAP20112AEA 386.83 EUR
ABGEAP20112A
Anti-RALYL Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RALYL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR