Tlačiť

Anti-ZAR1 Rabbit Polyclonal Antibody

Dodávateľ: Abgent
ABGEAP16486CEA 387.67 EUR
ABGEAP16486C
Anti-ZAR1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ZAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR