Tlačiť

Anti-SDCCAG8 Rabbit Polyclonal Antibody

Dodávateľ: Abgent
ABGEAP17462AEA 386.83 EUR
ABGEAP17462A
Anti-SDCCAG8 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SDCCAG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR