Tlačiť
Volumetric flasks, Class A, USP, glass, clear, BLAUBRAND®, NS plastic
Dodávateľ: Brand
BLAUBRAND®
BRNDUSP37249EA 72.46 EUR
BRNDUSP37249
Volumetric flasks, Class A, USP, glass, clear, BLAUBRAND®, NS plastic
Baňky Baňky odměrné
Borosilicate glass 3.3, clear with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
Certifikácie: DIN EN ISO 1042. Conformity certified (DE-M marking).
Informace o dodávce: Supplied with a batch number, batch certificate and with USP individual certificate.